2,2,4-Trimethylpentane, 99.5%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99.5%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99.5%, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99.5%, Thermo Scientific Chemicals
For HPLC
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA268810010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS37098-75-2
IUPAC Name5-(chlorosulfonyl)-2-fluorobenzoic acid
Molecular FormulaC7H4ClFO4S
InChI KeyCISMZCVQNVNWJW-UHFFFAOYSA-N
SMILESOC(=O)C1=C(F)C=CC(=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Titration ex Chloride)≥96.0%
Melting Point (clear melt)110.0-116.0?C
For HPLC
RUO – Research Use Only