1,4-Dioxane, 99.5%, for HPLC, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for HPLC, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for HPLC, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for HPLC, Thermo Scientific Chemicals
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA268340025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS319-03-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Formpowder
Assay (GC)> 97.5%
For HPLC
RUO – Research Use Only