2,2,4-Trimethylpentane, 99.5%, for analysis, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99.5%, for analysis, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99.5%, for analysis, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99.5%, for analysis, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Catalog number FSA265440010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS1070-34-4
IUPAC Name4-ethoxy-4-oxobutanoic acid
Molecular FormulaC6H10O4
InChI KeyLOLKAJARZKDJTD-UHFFFAOYSA-N
SMILESCCOC(=O)CCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)>94.0%
Appearance (Color)Colorless to yellow