γ-Undecalactone, 98%, Thermo Scientific Chemicals
γ-Undecalactone, 98%, Thermo Scientific Chemicals
γ-Undecalactone, 98%, Thermo Scientific Chemicals
γ-Undecalactone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
γ-Undecalactone, 98%, Thermo Scientific Chemicals
CAS: 104-67-6 | C11H20O2 | 184.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA259490250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS122-78-1
IUPAC Name2-phenylacetaldehyde
Molecular FormulaC8H8O
InChI KeyDTUQWGWMVIHBKE-UHFFFAOYSA-N
SMILESO=CCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5250-1.5350 @ 20°C
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥94.0%
Identification (FTIR)Conforms
Solution Test10% v/v in methanol: Clear