1-Amino-4-methylpiperazine, 99%, Thermo Scientific Chemicals
1-Amino-4-methylpiperazine, 99%, Thermo Scientific Chemicals
1-Amino-4-methylpiperazine, 99%, Thermo Scientific Chemicals
1-Amino-4-methylpiperazine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Amino-4-methylpiperazine, 99%, Thermo Scientific Chemicals
CAS: 6928-85-4 | C5H13N3 | 115.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA251390250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS123-41-1
IUPAC Name(2-hydroxyethyl)trimethylazanium hydroxide
Molecular FormulaC5H15NO2
InChI KeyKIZQNNOULOCVDM-UHFFFAOYSA-M
SMILES[OH-].C[N+](C)(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥44.0 to ≤48.0%
Appearance (Color)Colorless to brown
CommentMay turn hazy in appearance during storage. Any haziness should not affect it's function
FormViscous liquid
Assay (Non-aqueous acid-base Titration)≥44.0 to ≤48.0%