Sodium, 30wt% dispersion in toluene, <0.1 mm particle size
Sodium, 30wt% dispersion in toluene, &lt;0.1 mm particle size
Sodium, 30wt% dispersion in toluene, &lt;0.1 mm particle size
Sodium, 30wt% dispersion in toluene, &lt;0.1 mm particle size
Thermo Scientific Chemicals
Sodium, 30wt% dispersion in toluene, <0.1 mm particle size
CAS: 7440-23-5 | Na | 22.99 g/mol
Have Questions?
Catalog NumberQuantity
FSA243262500250 mL
Catalog number FSA243262500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Chemical Identifiers
CAS19961-27-4
IUPAC Nameethyl(propan-2-yl)amine
Molecular FormulaC5H13N
InChI KeyRIVIDPPYRINTTH-UHFFFAOYSA-N
SMILESCCNC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3895-1.3945 @ 20?C
Appearance (Color)Clear, colorless
FormLiquid
Assay (GC)>97.5%