Tetracycline hydrochloride, can be used as secondary standard
Tetracycline hydrochloride, can be used as secondary standard
Tetracycline hydrochloride, can be used as secondary standard
Tetracycline hydrochloride, can be used as secondary standard
Thermo Scientific Chemicals
Tetracycline hydrochloride, can be used as secondary standard
Tetracycline hydrochloride, CAS # 64-75-5, is a wide-spectrum antibiotic that targets both gram-negative and gram-positive bacteria.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2.5 g
Packaging:
Glass Bottle
Catalog number FSA233105000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetracycline hydrochloride
TypeTetracycline Hydrochloride
Physical FormPowder
CAS64-75-5
Health Hazard 1LIGHT SENSITIVE
View more
General Description
  • Tetracycline hydrochloride is the hydrochloride salt of the antibiotic tetracycline
  • Tetracycline blocks amino acyl-tRNA binding during bacterial protein synthesis
  • Tetracycline is a broad-spectrum naphthacene antibiotic that inhibits growth of both gram positive and gram negative bacteria
Applications
  • Tetracycline may be used as a selection agent in cell culture
  • It may also be used to activate inducible gene expression in engineered cells
RUO – Research Use Only
General References:
  1. Chopra, I.; Roberts, M. Tetracycline antibiotics: mode of action, applications, molecular biology, and epidemiology of bacterial resistance. Microbiol Mol Biol Rev. 2001, 65(2), 232-60.
  2. Kenny, P.A.; Enver, T., Ashworth, A. Retroviral vectors for establishing tetracycline-regulated gene expression in an otherwise recalcitrant cell line. BMC Mol Biol. 2002, 3:13.