Taurocholic acid, sodium salt hydrate, 98%
Taurocholic acid, sodium salt hydrate, 98%
Taurocholic acid, sodium salt hydrate, 98%
Taurocholic acid, sodium salt hydrate, 98%
Taurocholic acid, sodium salt hydrate, 98%
Taurocholic acid, sodium salt hydrate, 98%
Taurocholic acid, sodium salt hydrate, 98%
Taurocholic acid, sodium salt hydrate, CAS # 312693-83-7, also known as sodium taurocholate hydrate, is a bile acid taurine conjugate form in the liver with detergent capabilities.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA230371000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS591-31-1
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤1%
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5515-1.5555 @ 20?C
General Description
  • Taurocholic acid, sodium salt hydrate is an anionic bile acid detergent synthesized from cholic acid
  • This compound shows a choleretic activity promoting the secretion of bile from the liver
Applications
  • Taurocholic acid is used in the emulsification of lipids/ fats
  • It is used for protein solubilization and in the preparation of digestive enzymes for research applications
  • It can be used to determine conjugated and unconjugated bile salts in serum and jejunal fluid
RUO – Research Use Only