D(+)-Biotin, 98%
D(+)-Biotin, 98%
D(+)-Biotin, 98%
Thermo Scientific Chemicals
D(+)-Biotin, 98%
CAS: 58-85-5 | C10H16N2O3S | 244.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 mg
1 g
5 g
Packaging:
View all
Glass bottle
Catalog number FSA230090250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS694-80-4
IUPAC Name1-bromo-2-chlorobenzene
Molecular FormulaC6H4BrCl
InChI KeyQBELEDRHMPMKHP-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
Assay (GC)≥98.0%
Refractive Index1.5795-1.5835 @ 20?C