3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,3',5,5'-Tetramethylbenzidine, 99+%, Thermo Scientific Chemicals
CAS: 54827-17-7 | C16H20N2 | 240.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA229280250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4559-70-0
IUPAC Name(phenylphosphoroso)benzene
Molecular FormulaC12H11OP
InChI KeyASUOLLHGALPRFK-UHFFFAOYSA-N
SMILESO=P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or granules
Assay (HPLC)≥96.0%
Melting Point (clear melt)48.0-58.0?C