1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals
1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals
1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals
1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, Thermo Scientific Chemicals
CAS: 6004-24-6 | C21H40ClNO | 358.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number FSA226991000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS51-55-8
IUPAC Name8-methyl-8-azabicyclo[3.2.1]octan-3-yl 3-hydroxy-2-phenylpropanoate
Molecular FormulaC17H23NO3
InChI KeyRKUNBYITZUJHSG-UHFFFAOYNA-N
SMILESCN1C2CCC1CC(C2)OC(=O)C(CO)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point114-118°C
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay from Supplier's CofA≥98.5
Water Content (Karl Fischer Titration)≤0.2%