L(+)-Potassium hydrogen tartrate, 99%, Thermo Scientific Chemicals
L(+)-Potassium hydrogen tartrate, 99%, Thermo Scientific Chemicals
L(+)-Potassium hydrogen tartrate, 99%, Thermo Scientific Chemicals
L(+)-Potassium hydrogen tartrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
L(+)-Potassium hydrogen tartrate, 99%, Thermo Scientific Chemicals
CAS: 868-14-4 | C4H5KO6 | 188.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA222672500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS30525-89-4
IUPAC Nameformaldehyde
Molecular FormulaCH2O
InChI KeyWSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILESC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (HCOOH)=<0.03 %
Appearance (Color)White
Appearance (Form)Powder or beads
Infrared spectrumConforms
Titration with HCl>=95 %