Strontium nitrate, 99+%, for analysis, Thermo Scientific Chemicals
Strontium nitrate, 99+%, for analysis, Thermo Scientific Chemicals
Strontium nitrate, 99+%, for analysis, Thermo Scientific Chemicals
Strontium nitrate, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Strontium nitrate, 99+%, for analysis, Thermo Scientific Chemicals
Strontium nitrate, 99+%, N2O6Sr, CAS Number-10042-76-9, bdg873aqzl, strontium dinitrate, nitric acid, strontium salt 2:1, hsdb 787, nitric acid, strontium salt, nitrate de strontium french, strontium nitrate sr no3 2, strontium nitrate, strontium ii nitrate 1:2, unii-bdg873aqzl, 1kg
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA222562500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS103-71-9
IUPAC Nameisocyanatobenzene
Molecular FormulaC7H5NO
InChI KeyDGTNSSLYPYDJGL-UHFFFAOYSA-N
SMILESO=C=NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.5350 to 1.5380 (20°C, 589 nm)