Dimethyl fumarate, 99%, Thermo Scientific Chemicals
Dimethyl fumarate, 99%, Thermo Scientific Chemicals
Dimethyl fumarate, 99%, Thermo Scientific Chemicals
Dimethyl fumarate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethyl fumarate, 99%, Thermo Scientific Chemicals
CAS: 624-49-7 | C6H8O4 | 144.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA222185000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDimethyl fumarate
CAS624-49-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/e