2-Chloroethyl isocyanate, 98%, Thermo Scientific Chemicals
2-Chloroethyl isocyanate, 98%, Thermo Scientific Chemicals
2-Chloroethyl isocyanate, 98%, Thermo Scientific Chemicals
2-Chloroethyl isocyanate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chloroethyl isocyanate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA221190010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Chloroethyl isocyanate
Name Note98%
CAS1943-83-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger