Cobalt(II) nitrate hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Cobalt(II) nitrate hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Cobalt(II) nitrate hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Cobalt(II) nitrate hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cobalt(II) nitrate hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals
CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA219215000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS12027-06-4
IUPAC Nameammonium iodide
Molecular FormulaH4IN
InChI KeyUKFWSNCTAHXBQN-UHFFFAOYSA-N
SMILES[NH4+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals
Appearance (Color)White
Assay (special)Iodine: 85.5-89.5%
Total Metal Impurities≤0.005% (50 ppm)
For analysis
RUO – Research Use Only