Cyclohexyl iodide, 98%, stabilized, Thermo Scientific Chemicals
Cyclohexyl iodide, 98%, stabilized, Thermo Scientific Chemicals
Cyclohexyl iodide, 98%, stabilized, Thermo Scientific Chemicals
Cyclohexyl iodide, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexyl iodide, 98%, stabilized, Thermo Scientific Chemicals
CAS: 626-62-0 | C6H11I | 210.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA218490250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16029-98-4
IUPAC Nameiodotrimethylsilane
Molecular FormulaC3H9ISi
InChI KeyCSRZQMIRAZTJOY-UHFFFAOYSA-N
SMILESC[Si](C)(C)I
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4700 to 1.4800 (20°C, 589 nm)
Stabilizercopper
Appearance (Color)Clear colorless to red