Acetic acid, ammonium salt, 98+%, for analysis, Thermo Scientific Chemicals
Acetic acid, ammonium salt, 98+%, for analysis, Thermo Scientific Chemicals
Acetic acid, ammonium salt, 98+%, for analysis, Thermo Scientific Chemicals
Acetic acid, ammonium salt, 98+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, ammonium salt, 98+%, for analysis, Thermo Scientific Chemicals
CAS: 631-61-8 | C2H7NO2 | 77.083 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA218365000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS18323-96-1
IUPAC Nameytterbium(3+) tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-dioxooctan-4-ide)
Molecular FormulaC30H30F21O6Yb
InChI KeyQYXREIJIWYJRCU-UHFFFAOYSA-N
SMILES[Yb+3].CC(C)(C)C(=O)[CH-]C(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(=O)[CH-]C(=O)C(F)(F)C(F)(F)C(F)(F)F.CC(C)(C)C(=O)[CH-]C(=O)C(F)(F)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Appearance (Color)Pale yellow
Chloride content< 0.1%
Assay (unspecified)> 98.5%
Biological buffer for analysis
RUO – Research Use Only