4-Acetophenetidide, 97%, Thermo Scientific Chemicals
4-Acetophenetidide, 97%, Thermo Scientific Chemicals
4-Acetophenetidide, 97%, Thermo Scientific Chemicals
4-Acetophenetidide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Acetophenetidide, 97%, Thermo Scientific Chemicals
CAS: 62-44-2 | C10H13NO2 | 179.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA216591000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS498-66-8
IUPAC Namebicyclo[2.2.1]hept-2-ene
Molecular FormulaC7H10
InChI KeyJFNLZVQOOSMTJK-UHFFFAOYNA-N
SMILESC1CC2CC1C=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Stabilizer0.1 % phenolic antioxidant
Appearance (Color)White
Infrared spectrumConforms
Melting point42°C to 46°C
GC>=98.5 %