Acetonitrile-d3, for NMR, contains 1 v/v% TMS, 99 atom % D, Thermo Scientific Chemicals
Acetonitrile-d3, for NMR, contains 1 v/v% TMS, 99 atom % D, Thermo Scientific Chemicals
Acetonitrile-d3, for NMR, contains 1 v/v% TMS, 99 atom % D, Thermo Scientific Chemicals
Acetonitrile-d3, for NMR, contains 1 v/v% TMS, 99 atom % D, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetonitrile-d3, for NMR, contains 1 v/v% TMS, 99 atom % D, Thermo Scientific Chemicals
CAS: 2206-26-0 | C2H3N | 44.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
25 mL
Packaging:
Glass Bottle
Catalog number FSA214530500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS13918-92-8
IUPAC Name2,4-difluorobenzene-1-sulfonyl chloride
Molecular FormulaC6H3ClF2O2S
InChI KeyFJSAJUXIHJIAMD-UHFFFAOYSA-N
SMILESFC1=CC(F)=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5155-1.5195 @ 20?C
Appearance (Color)Clear colorless to yellow
For NMR
RUO – Research Use Only