Cobalt(II) chloride, 97%, anhydrous, Thermo Scientific Chemicals
Cobalt(II) chloride, 97%, anhydrous, Thermo Scientific Chemicals
Cobalt(II) chloride, 97%, anhydrous, Thermo Scientific Chemicals
Cobalt(II) chloride, 97%, anhydrous, Thermo Scientific Chemicals
Cobalt(II) chloride, 97%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cobalt(II) chloride, 97%, anhydrous, Thermo Scientific Chemicals
CAS: 7646-79-9 | Cl2Co | 129.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number FSA214135000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Boiling Point1049.0°C
Chemical Name or MaterialCobalt(II) chloride
Melting Point735.0°C
Name Note97%
CAS7646-79-9
View more
Suitable for battery materials development.
RUO – Research Use Only