3-Aminoquinoline, 99%, Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
CAS: 580-17-6 | C9H8N2 | 144.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA213780050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS630-10-4
IUPAC Name[(C-selanylcarbonimidoyl)amino]-λ¹-selanyl
Molecular FormulaCH3N2Se
InChI KeyZUCFIOXGLBMWHV-UHFFFAOYSA-N
SMILESNC([Se])=N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder
Appearance (Color)Pale cream to grey to violet
Assay (unspecified)≥63.2% Se (equivalent to ≥98.5% purity)