Acetic acid, sodium salt, 99%, for biochemistry, Anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for biochemistry, Anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for biochemistry, Anhydrous, Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for biochemistry, Anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, sodium salt, 99%, for biochemistry, Anhydrous, Thermo Scientific Chemicals
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Catalog number FSA212662500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS107-15-3
IUPAC Nameethane-1,2-diamine
Molecular FormulaC2H8N2
InChI KeyPIICEJLVQHRZGT-UHFFFAOYSA-N
SMILESNCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.5 % (Coulometric)
Appearance (Form)Liquid
Refractive index1.4555 to 1.4575 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
View more
Biological buffer
RUO – Research Use Only