Naphthol AS, 99%
Naphthol AS, 99%
Naphthol AS, 99%
Thermo Scientific Chemicals
Naphthol AS, 99%
CAS: 92-77-3 | C17H13NO2 | 263.28 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA212181000100 gGlass bottle
Catalog number FSA212181000
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13963-57-0
IUPAC Namealuminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC15H21AlO6
InChI KeyKILURZWTCGSYRE-LNTINUHCSA-K
SMILES[Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Melting point190°C to 195°C
Appearance (Form)Crystalline powder
Titration Complexometric>=96.0 %