N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
10 L
25 L
Packaging:
View all
Glass Bottle
Catalog number FSA210585000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS21548-73-2
IUPAC Namedisilver(1+) sulfanediide
Molecular FormulaAg2S
InChI KeyXUARKZBEFFVFRG-UHFFFAOYSA-N
SMILES[S--].[Ag+].[Ag+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark grey to black
FormPowder
Assay from Suppliers CofA≥98.5%
For analysis
RUO – Research Use Only