CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
CHES, 98+%, for biochemistry
Thermo Scientific Chemicals
CHES, 98+%, for biochemistry
CAS: 103-47-9 | C8H17NO3S | 207.288 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Plastic bottle
Catalog number FSA208181000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS121-72-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow or pale brown
FormLiquid
Assay (GC)≥97.0%
Refractive Index1.5475-1.5525 @ 20?C
Biological buffer
RUO – Research Use Only