Formamide, 99.5%, for analysis, Thermo Scientific Chemicals
Formamide, 99.5%, for analysis, Thermo Scientific Chemicals
Formamide, 99.5%, for analysis, Thermo Scientific Chemicals
Formamide, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Formamide, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 75-12-7 | CH3NO | 45.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Catalog number FSA205820010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS100-48-1
IUPAC Namepyridine-4-carbonitrile
Molecular FormulaC6H4N2
InChI KeyGPHQHTOMRSGBNZ-UHFFFAOYSA-N
SMILESN#CC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige to light brown
Appearance (Form)Powder or crystals or flakes
Melting point76°C to 81°C
Infrared spectrumConforms
GC>=97.5 %
For analysis
RUO – Research Use Only