Florisil™, 100-200 mesh
Florisil™, 100-200 mesh
Florisil™, 100-200 mesh
Florisil™, 100-200 mesh
Thermo Scientific Chemicals
Florisil™, 100-200 mesh
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
5 kg
Catalog number FSA205442500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS372-47-4
IUPAC Name3-fluoropyridine
Molecular FormulaC5H4FN
InChI KeyCELKOWQJPVJKIL-UHFFFAOYSA-N
SMILESFC1=CN=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless to yellow
Refractive index1.4720 to 1.4750 (20°C, 589 nm)