Florisil™, 100-200 mesh
Florisil™, 100-200 mesh
Florisil™, 100-200 mesh
Florisil™, 100-200 mesh
Thermo Scientific Chemicals
Florisil™, 100-200 mesh
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
500 g
Catalog number FSA205440050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Request bulk or custom format
Chemical Identifiers
CAS98-51-1
IUPAC Name1-tert-butyl-4-methylbenzene
Molecular FormulaC11H16
InChI KeyQCWXDVFBZVHKLV-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.4910 to 1.4930 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=95 %