Calcium, 99%, granular, Thermo Scientific Chemicals
Calcium, 99%, granular, Thermo Scientific Chemicals
Calcium, 99%, granular, Thermo Scientific Chemicals
Calcium, 99%, granular, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calcium, 99%, granular, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic Bottle
Catalog number FSA201181000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS20312-36-1
IUPAC Name(2S)-2-hydroxy-3-phenylpropanoate
Molecular FormulaC9H9O3
InChI KeyVOXXWSYKYCBWHO-QMMMGPOBSA-M
SMILESO[C@@H](CC1=CC=CC=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale yellow
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)121.0-127.0?C
Optical Rotation-22.0 ± 2.0? (C = 1 in water)