Copper(I) chloride, 97%, pure, Thermo Scientific Chemicals
Copper(I) chloride, 97%, pure, Thermo Scientific Chemicals
Copper(I) chloride, 97%, pure, Thermo Scientific Chemicals
Copper(I) chloride, 97%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper(I) chloride, 97%, pure, Thermo Scientific Chemicals
CAS: 7758-89-6 | ClCu | 99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA199645000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Boiling Point1490.0°C
Chemical Name or MaterialCopper(I) chloride
Melting Point430.0°C
Name Note95%
CAS7758-89-6