Copper(I) bromide, 98%, extra pure, Thermo Scientific Chemicals
Copper(I) bromide, 98%, extra pure, Thermo Scientific Chemicals
Copper(I) bromide, 98%, extra pure, Thermo Scientific Chemicals
Copper(I) bromide, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper(I) bromide, 98%, extra pure, Thermo Scientific Chemicals
CAS: 7787-70-4 | BrCu | 143.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number FSA199582500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point1345.0°C
Chemical Name or MaterialCopper(I) bromide
Melting Point504.0°C
CAS7787-70-4
ColorGreen