Phenylbutazone, 99+%
Phenylbutazone, 99+%
Phenylbutazone, 99+%
Phenylbutazone, 99+%
Phenylbutazone, 99+%
CAS: 50-33-9 | C19H20N2O2 | 308.38 g/mol
Have Questions?
Catalog number FSA199310050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS207801-28-3
IUPAC Nameyttrium(3+) triacetate
Molecular FormulaC6H9O6Y
InChI KeyJUWHRRAPBUAYTA-UHFFFAOYSA-K
SMILES[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Total conc. of imp.as REO=<0.1 %
Appearance (Color)White
Appearance (Form)Crystalline powder and/or lumps