Ruthenium, 99.9%, (trace metal basis), -200 mesh, powder, Thermo Scientific Chemicals
Ruthenium, 99.9%, (trace metal basis), -200 mesh, powder, Thermo Scientific Chemicals
Ruthenium, 99.9%, (trace metal basis), -200 mesh, powder, Thermo Scientific Chemicals
Ruthenium, 99.9%, (trace metal basis), -200 mesh, powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ruthenium, 99.9%, (trace metal basis), -200 mesh, powder, Thermo Scientific Chemicals
CAS: 7440-18-8 | Ru | 101.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA198100010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS35132-20-8
IUPAC Name(1R,2R)-1,2-diphenylethane-1,2-bis(aminium)
Molecular FormulaC14H18N2
InChI KeyPONXTPCRRASWKW-ZIAGYGMSSA-P
SMILES[NH3+][C@@H]([C@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
GC>=98.0 %
Appearance (Form)Powder or crystalline powder or crystals
Specific optical rotation+101° to +107° (20°C, 589 nm) (C=1, Methanol)
View more
for ultra trace analysis
RUO – Research Use Only