4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromobiphenyl, 98+%, Thermo Scientific Chemicals
CAS: 92-66-0 | C12H9Br | 233.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA197981000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6296-99-7
IUPAC Name1,3-diethyl 2-(aminomethylidene)propanedioate
Molecular FormulaC8H13NO4
InChI KeyWDTWMEZMHUEZKH-UHFFFAOYSA-N
SMILESCCOC(=O)C(=CN)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White
FormCrystals or powder or crystalline powder