Iron, 99%, powder, -70 mesh (<212 micron), Thermo Scientific Chemicals
Iron, 99%, powder, -70 mesh (&lt;212 micron), Thermo Scientific Chemicals
Iron, 99%, powder, -70 mesh (&lt;212 micron), Thermo Scientific Chemicals
Iron, 99%, powder, -70 mesh (&lt;212 micron), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iron, 99%, powder, -70 mesh (<212 micron), Thermo Scientific Chemicals
CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
10 kg
Packaging:
Plastic bottle
Catalog number FSA197815000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery white
FormWire, 0.5mm (0.02in) diameter, hardened.
Assay from Supplier's CofA≥99.999% (metals basis)
Reduction reagent
RUO – Research Use Only