Aluminum sulfate, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Aluminum sulfate, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aluminum sulfate, 99.999%, (trace metal basis), extra pure, Thermo Scientific Chemicals
CAS: 10043-01-3 | Al2O12S3 | 342.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA192431000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10022-31-8
IUPAC Namebarium(2+) dinitrate
Molecular FormulaBaN2O6
InChI KeyIWOUKMZUPDVPGQ-UHFFFAOYSA-N
SMILES[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
Complexometric Titration≥98.5 to ≤101.5%
For ultra trace analysis
RUO – Research Use Only