Cobalt(II) chloride hexahydrate, for analysis, Thermo Scientific Chemicals
Cobalt(II) chloride hexahydrate, for analysis, Thermo Scientific Chemicals
Cobalt(II) chloride hexahydrate, for analysis, Thermo Scientific Chemicals
Cobalt(II) chloride hexahydrate, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cobalt(II) chloride hexahydrate, for analysis, Thermo Scientific Chemicals
CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA192092500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS50-01-1
IUPAC Namehydrogen guanidine chloride
Molecular FormulaCH6ClN3
InChI KeyPJJJBBJSCAKJQF-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms to structure and standard
Identification APasses test
UVat 260 nm A: =<0.030
Appearance (Color)Colorless to white
Titration with HClO4>=99.5 %