m-Xylene, 99+%, extra pure, Thermo Scientific Chemicals
m-Xylene, 99+%, extra pure, Thermo Scientific Chemicals
m-Xylene, 99+%, extra pure, Thermo Scientific Chemicals
m-Xylene, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
m-Xylene, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 108-38-3 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
Catalog number FSA180862500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Materialm-Xylene
Melting Point-48.0°C
CAS108-38-3
CAS Min %99.0