Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Terephthalic acid, 99+%, Thermo Scientific Chemicals
CAS: 100-21-0 | C8H6O4 | 166.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA180725000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS490-78-8
IUPAC Name1-(2,5-dihydroxyphenyl)ethan-1-one
Molecular FormulaC8H8O3
InChI KeyWLDWSGZHNBANIO-UHFFFAOYSA-N
SMILESCC(=O)C1=CC(O)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point203°C to 208°C (Dec)
HPLC>=96.0 %