Phenylarsine oxide, 97%, Thermo Scientific Chemicals
Phenylarsine oxide, 97%, Thermo Scientific Chemicals
Phenylarsine oxide, 97%, Thermo Scientific Chemicals
Phenylarsine oxide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenylarsine oxide, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA180350250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS554-13-2
IUPAC Namedilithium(1+) carbonate
Molecular FormulaCLi2O3
InChI KeyXGZVUEUWXADBQD-UHFFFAOYSA-L
SMILES[Li+].[Li+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Total trace metal impurities=<0.001 %