tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals
CAS: 75-91-2 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
250 g
500 g
Packaging:
Plastic bottle
Catalog number FSA180340025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Materialtert-Butyl hydroperoxide
CAS75-91-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
CAS7732-18-5