tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
2.5 L
Packaging:
Glass Bottle
Catalog number FSA177042500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS873-51-8
IUPAC Namedichloro(phenyl)borane
Molecular FormulaC6H5BCl2
InChI KeyNCQDQONETMHUMY-UHFFFAOYSA-N
SMILESClB(Cl)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5450 to 1.5470 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration Argentometric96.0 to 104.0 % (ex Cl)