Acetone-d6, for NMR, +99% atom D, Thermo Scientific Chemicals
Acetone-d6, for NMR, +99% atom D, Thermo Scientific Chemicals
Acetone-d6, for NMR, +99% atom D, Thermo Scientific Chemicals
Acetone-d6, for NMR, +99% atom D, Thermo Scientific Chemicals
Acetone-d6, for NMR, +99% atom D, Thermo Scientific Chemicals
CAS: 666-52-4 | C3H6O | 64.117 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
Catalog number FSA174900500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Request bulk or custom format
Chemical Identifiers
CAS380430-57-9
IUPAC Name(4-methanesulfonamidophenyl)boronic acid
Molecular FormulaC7H10BNO4S
InChI KeyNDVJJEADFLTFCD-UHFFFAOYSA-N
SMILESCS(=O)(=O)NC1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Additional infoMay contain varying amounts of anhydrides
Appearance (Color)Off-white to cream to light brown
NMR (1H-NMR)Consistent with structure
Appearance (Form)Solid
View more
For NMR
RUO – Research Use Only