(3-Mercaptopropyl)trimethoxysilane, 85%, Tech., Thermo Scientific Chemicals
(3-Mercaptopropyl)trimethoxysilane, 85%, Tech., Thermo Scientific Chemicals
(3-Mercaptopropyl)trimethoxysilane, 85%, Tech., Thermo Scientific Chemicals
(3-Mercaptopropyl)trimethoxysilane, 85%, Tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals
(3-Mercaptopropyl)trimethoxysilane, 85%, Tech., Thermo Scientific Chemicals
CAS: 4420-74-0 | C6H16O3SSi | 196.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number FSA174652500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS420-37-1
IUPAC Nametetrafluoroboranuide; trimethyloxidanium
Molecular FormulaC3H9BF4O
InChI KeyCZVZBKHWOFJNCR-UHFFFAOYSA-N
SMILESC[O+](C)C.F[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
Titration with NaOH94.0 to 106.0 %
Appearance (Color)White to off-white