Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Activated charcoal, NORIT(R) SA 2, decolorizing, Thermo Scientific Chemicals
CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic drum
Catalog number FSA171530010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS1724-39-6
IUPAC Namecyclododecanol
Molecular FormulaC12H24O
InChI KeySFVWPXMPRCIVOK-UHFFFAOYSA-N
SMILESOC1CCCCCCCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or fused/lumpy solid
Appearance (Color)White
Assay (GC)≥98.5%
Melting Point (clear melt)75.0-81.0?C
Filter aid
RUO – Research Use Only