Primaquine diphosphate, 98%
Primaquine diphosphate, 98%
Primaquine diphosphate, 98%
Primaquine diphosphate, 98%
Thermo Scientific Chemicals
Primaquine diphosphate, 98%
CAS: 63-45-6 | C15H21N32H3PO4 | 455.34 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA1708700101 gGlass bottle
Catalog number FSA170870010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS28240-69-9
IUPAC Namedichloro[2-(dichlorophosphanyl)ethyl]phosphane
Molecular FormulaC2H4Cl4P2
InChI KeySBWAJHLQMFBNIN-UHFFFAOYSA-N
SMILESClP(Cl)CCP(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay by NMR≥95.0%