2-Bromoanisole, 97%, Thermo Scientific Chemicals
2-Bromoanisole, 97%, Thermo Scientific Chemicals
2-Bromoanisole, 97%, Thermo Scientific Chemicals
2-Bromoanisole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromoanisole, 97%, Thermo Scientific Chemicals
CAS: 578-57-4 | C7H7BrO | 187.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA170261000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS17551-52-9
IUPAC Name6-chloro-2-methyl-1,4-dihydropyrimidin-4-one
Molecular FormulaC5H5ClN2O
InChI KeyQNANRGHPXMUJQC-UHFFFAOYSA-N
SMILESCC1=NC(=O)C=C(Cl)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
FormPowder
Appearance (Color)White
Melting Point (clear melt)228.0-238.0?C