Acetic acid, ammonium salt, 98%, pure, Thermo Scientific Chemicals
Acetic acid, ammonium salt, 98%, pure, Thermo Scientific Chemicals
Acetic acid, ammonium salt, 98%, pure, Thermo Scientific Chemicals
Acetic acid, ammonium salt, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetic acid, ammonium salt, 98%, pure, Thermo Scientific Chemicals
CAS: 631-61-8 | C2H7NO2 | 77.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic Bottle
Catalog number FSA169880010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetic acid, ammonium salt
CAS631-61-8
Melting Point114.0°C
ColorWhite
EINECS Number211-162-9