Heptafluorobutyric anhydride, 98%, Thermo Scientific Chemicals
Heptafluorobutyric anhydride, 98%, Thermo Scientific Chemicals
Heptafluorobutyric anhydride, 98%, Thermo Scientific Chemicals
Heptafluorobutyric anhydride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Heptafluorobutyric anhydride, 98%, Thermo Scientific Chemicals
CAS: 336-59-4 | C8F14O3 | 410.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA169261000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6132-02-1
IUPAC Namedisodium decahydrate carbonate
Molecular FormulaCH20Na2O13
InChI KeyXYQRXRFVKUPBQN-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals
Titration with HCl99.0 to 102.0 %
Chloride (Cl)=<10 ppm
Sulfate (SO4)=<30 ppm
View more
Acylation reagent
RUO – Research Use Only